Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloro[2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]palladium(II)

Chemical Identifiers

Chemical Name or Material Dichloro[2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]palladium(II)
CAS 253157-79-8
Formula Weight 800.01
MDL Number MFCD00075254
Synonym PdCl2(BINAP); [2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl]dichloropalladium(II)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4658803

  SDS
alfa aesar™
4658803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4658806

  SDS
alfa aesar™
4658806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 230° to 300°C (dec.)
Formula Weight 800.01
CAS 253157-79-8
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Color Orange
Synonym PdCl2(BINAP); [2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl]dichloropalladium(II)
Health Hazard 1 Warning
Chemical Name or Material Dichloro[2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]palladium(II)
Physical Form Crystalline Solid
MDL Number MFCD00075254
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.