missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichloro[2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]palladium(II), Alfa Aesar™

Manufacturer:  Alfa Aesar 4658806

 View more versions of this product

Catalog No. AA4658806



Specifications

Specifications

Dichloro[2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]palladium(II)
230°C to 300°C (decomposition)
253157-79-8
MFCD00075254
VDHAUMFISVWIRX-UHFFFAOYSA-L
[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;palladium(2+);dichloride
25171195
Crystalline Solid
Orange
5g
C44H32Cl2P2Pd
r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl palladium ii chloride, 2,2'-bis diphenylphosphino-1,1'-binaphthylpalladium ii chloride, s-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl palladium ii chloride, pd r-binap cl2, palladium 2+ binap dichloride, dichloro 2,2'-bis diphenylphosphino-1,1'-binaphthyly palladium ii, dichloro r-2,2'-bis diphenylphosphino-1,1'-binaphthyl palladium ii, dichloro s-?-2,2'-bis diphenylphosphino-1,1'-binaphthyl palladium ii, palladium, 1s-1,1'-binaphthalene-2,2'-diylbis diphenylphosphine-kp dichloro-, sp-4-2
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.[Cl-].[Cl-].[Pd+2]
800.008
800.01

Chemical Identifiers

253157-79-8
800.008
VDHAUMFISVWIRX-UHFFFAOYSA-L
25171195
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.[Cl-].[Cl-].[Pd+2]
C44H32Cl2P2Pd
MFCD00075254
r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl palladium ii chloride, 2,2'-bis diphenylphosphino-1,1'-binaphthylpalladium ii chloride, s-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl palladium ii chloride, pd r-binap cl2, palladium 2+ binap dichloride, dichloro 2,2'-bis diphenylphosphino-1,1'-binaphthyly palladium ii, dichloro r-2,2'-bis diphenylphosphino-1,1'-binaphthyl palladium ii, dichloro s-?-2,2'-bis diphenylphosphino-1,1'-binaphthyl palladium ii, palladium, 1s-1,1'-binaphthalene-2,2'-diylbis diphenylphosphine-kp dichloro-, sp-4-2
[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;palladium(2+);dichloride
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents