missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloro(1,5-cyclooctadiene)platinum(II), Pt 51.6-52.6%

Manufacturer:  Alfa Aesar™ 1273901

 View more versions of this product

Catalog No. AA1273901



Specifications

Specifications

Dichloro(1,5-cyclooctadiene)platinum(II)
250°C (decomposition)
12080-32-9
MFCD00012413
Pt 51.6 to 52.6%
VVAOPCKKNIUEEU-PHFPKPIQSA-L
(1Z,5Z)-cycloocta-1,5-diene;platinum(2+);dichloride
6436378
Solid
Hygroscopic
0.25g
C8H12Cl2Pt
dichloro 1,5-cyclooctadiene platinum ii, 1,5-cyclooctadienedichloroplatinum, dichloro 1,5-cyclooctadiene platinum, platinum, dichloro 1,5-cyclooctadiene, 1z,5z-cycloocta-1,5-diene; platinum 2+ ; dichloride, platinum, dichloro 1,2,5,6-eta-1,5-cyclooctadiene, 1,5-cyclooctadieneplatinum ii dichloride, dichloro 1,5-cyclooctadiene platinum ii 250mg, dichloro 1,2,5,6-eta-cycloocta-1,5-diene platinum, platinum 2+ 1,5-cyclooctadiene, z,z-dichloride
Soluble in dichloromethane
C1CC=CCCC=C1.[Cl-].[Cl-].[Pt+2]
374.168
374.18

Chemical Identifiers

12080-32-9
374.168
VVAOPCKKNIUEEU-PHFPKPIQSA-L
6436378
C1CC=CCCC=C1.[Cl-].[Cl-].[Pt+2]
C8H12Cl2Pt
MFCD00012413
dichloro 1,5-cyclooctadiene platinum ii, 1,5-cyclooctadienedichloroplatinum, dichloro 1,5-cyclooctadiene platinum, platinum, dichloro 1,5-cyclooctadiene, 1z,5z-cycloocta-1,5-diene; platinum 2+ ; dichloride, platinum, dichloro 1,2,5,6-eta-1,5-cyclooctadiene, 1,5-cyclooctadieneplatinum ii dichloride, dichloro 1,5-cyclooctadiene platinum ii 250mg, dichloro 1,2,5,6-eta-cycloocta-1,5-diene platinum, platinum 2+ 1,5-cyclooctadiene, z,z-dichloride
(1Z,5Z)-cycloocta-1,5-diene;platinum(2+);dichloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents