missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloro(1,5-cyclooctadiene)palladium(II), Pd 36.7%

Manufacturer:  Alfa Aesar™ 1049303

 View more versions of this product

Catalog No. AA1049303



Specifications

Specifications

Dichloro(1,5-cyclooctadiene)palladium(II)
Yellow
12107-56-1
MFCD00012412
Pd 36.7%
RRHPTXZOMDSKRS-PHFPKPIQSA-L
(1Z,5Z)-cycloocta-1,5-diene;palladium(2+);dichloride
6436380
Crystalline
Hygroscopic
1g
C8H12Cl2Pd
dichloro 1,5-cyclooctadiene palladium ii, cycloocta-1,5-diene dichloropalladium, 1,5-cyclooctadienepalladium ii dichloride, 1z,5z-cycloocta-1,5-diene; palladium 2+ ; dichloride, dichloro 1,5-cyclooctadiene palladium, palladium, dichloro 1,5-cyclooctadiene, dichloro 1,2,5,6-eta-cycloocta-1,5-diene palladium, palladium, dichloro 1,2,5,6-eta-1,5-cyclooctadiene, dichloro 1,5-cyclooctadiene palladium ii ,, dichloro 1,5-cyclooctadiene palladium ii 1g
Soluble in dichloromethane
C1CC=CCCC=C1.[Cl-].[Cl-].[Pd+2]
285.504
285.49

Chemical Identifiers

12107-56-1
285.504
RRHPTXZOMDSKRS-PHFPKPIQSA-L
6436380
C1CC=CCCC=C1.[Cl-].[Cl-].[Pd+2]
C8H12Cl2Pd
MFCD00012412
dichloro 1,5-cyclooctadiene palladium ii, cycloocta-1,5-diene dichloropalladium, 1,5-cyclooctadienepalladium ii dichloride, 1z,5z-cycloocta-1,5-diene; palladium 2+ ; dichloride, dichloro 1,5-cyclooctadiene palladium, palladium, dichloro 1,5-cyclooctadiene, dichloro 1,2,5,6-eta-cycloocta-1,5-diene palladium, palladium, dichloro 1,2,5,6-eta-1,5-cyclooctadiene, dichloro 1,5-cyclooctadiene palladium ii ,, dichloro 1,5-cyclooctadiene palladium ii 1g
(1Z,5Z)-cycloocta-1,5-diene;palladium(2+);dichloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents