Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloro[1,2-bis(diphenylphosphino)ethane]palladium(II), Pd 18.5%

Chemical Identifiers

Chemical Name or Material Dichloro[1,2-bis(diphenylphosphino)ethane]palladium(II)
CAS 19978-61-1
Linear Formula PdCl2[(C6H5)2PCH2CH2P(C6H5)2]
Formula Weight 575.75
MDL Number MFCD00015702
Synonym [1,2-Bis(diphenylphosphino)ethane]dichloropalladium(II); [1,2-Bis(diphenylphosphino)ethane]palladium(II) chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1877904

  SDS
alfa aesar™
1877904
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1877909

  SDS
alfa aesar™
1877909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 575.75
Chemical Name or Material Dichloro[1,2-bis(diphenylphosphino)ethane]palladium(II)
Physical Form Powder
Linear Formula PdCl2[(C6H5)2PCH2CH2P(C6H5)2]
MDL Number MFCD00015702
Color Dark yellow
CAS 19978-61-1
Melting Point >360°C
Synonym [1,2-Bis(diphenylphosphino)ethane]dichloropalladium(II); [1,2-Bis(diphenylphosphino)ethane]palladium(II) chloride
Concentration Pd 18.5%
Sensitivity Hygroscopic
Quantity 2g