Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), Pd 13.5-15.5%

Chemical Identifiers

Chemical Name or Material Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II)
CAS 72287-26-4
Formula Weight 731.72
MDL Number MFCD00015757
Synonym [1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4650303

  SDS
alfa aesar™
4650303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II)
Concentration Pd 13.5 to 15.5%
Color Orange
Melting Point 266° to 283°C
Quantity 1g
CAS 72287-26-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Solid
Formula Weight 731.72
MDL Number MFCD00015757
Synonym [1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.