Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II), 99%

$94.93 - $317.45

Chemical Identifiers

Chemical Name or Material Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II)
CAS 67292-34-6
Molecular Formula C34H28Cl2FeNiP2
Formula Weight 684.00g/mol
MDL Number MFCD00270284
Synonym [1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride; NiCl2(dppf)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2689503

 
alfa aesar™
H26895-03
1g Each for $94.93

AAH2689506

 
alfa aesar™
H26895-06
5g Each for $317.45
Description & Specifications

Specifications

Melting Point 285°C
Quantity 1g
Chemical Name or Material Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II)
CAS 67292-34-6
Assay 99%
Molecular Formula C34H28Cl2FeNiP2
Formula Weight 684.00g/mol
MDL Number MFCD00270284
Synonym [1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride; NiCl2(dppf)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H351-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P342+P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
diphosphine 1,1′-bis(diphenylphosphino)ferrocene (dppf) are used in transition metal-catalyzed reactions to synthesize pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents

Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P342+P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H351-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
May cause an allergic skin reaction.