Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloro[1,1'-bis(diphenylphosphino)ferrocene]cobalt(II), 98%

$93.27 - $344.80

Chemical Identifiers

Chemical Name or Material Dichloro[1,1'-bis (diphenylphosphino)ferrocene]cobalt (II)
CAS 67292-36-8
Molecular Formula C34H28Cl2FeP2Co
Formula Weight 684.23
MDL Number MFCD03427161
Synonym [1,1'-Bis(diphenylphosphino)ferrocene]cobalt(II) chloride; CoCl2(dppf)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4427406

 
alfa aesar™
4427406
5g Each for $344.80

AA4427403

 
alfa aesar™
4427403
1g Each for $93.27
Description & Specifications

Specifications

Formula Weight 684.23
Chemical Name or Material Dichloro[1,1'-bis (diphenylphosphino)ferrocene]cobalt (II)
Physical Form Powder
Assay 98%
Molecular Formula C34H28Cl2FeP2Co
MDL Number MFCD03427161
CAS 67292-36-8
Synonym [1,1'-Bis(diphenylphosphino)ferrocene]cobalt(II) chloride; CoCl2(dppf)
Color Dark green
Melting Point 224°C
Health Hazard 1 Warning
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.