missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichloro[1,1'-bis(diisopropylphosphino)ferrocene]palladium(II), 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar 4497803

 View more versions of this product

Catalog No. AA4497803



Specifications

Specifications

Dichloro[1,1'-bis (diisopropylphosphino)ferrocene]palladium (II)
282°C to 287°C
215788-65-1
MFCD11656082
595.65
98%
Orange-Red
1g
C22H36Cl2FeP2Pd
[1,1'-Bis(diisopropylphosphino)ferrocene]palladium(II) chloride; PdCl2(dippf)
Powder

Chemical Identifiers

215788-65-1
C22H36Cl2FeP2Pd
MFCD11656082
[1,1'-Bis(diisopropylphosphino)ferrocene]palladium(II) chloride; PdCl2(dippf)
Safety and Handling

Safety and Handling

Recommended Storage : 2°C to 8°C

SDS Documents