Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloro[1,1'-bis(dicyclohexylphosphino)ferrocene]palladium(II), Pd 14.1%

Chemical Identifiers

Chemical Name or Material Dichloro[1,1'-bis(dicyclohexylphosphino)ferrocene]palladium(II)
CAS 917511-90-1
Formula Weight 755.91
MDL Number MFCD11656081
Synonym PdCl2(dcypf); [1,1'-Bis(dicyclohexylphosphino)ferrocene]dichloropalladium(II)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4638303

 
alfa aesar™
4638303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 755.91
Melting Point 294° to 300°C
Chemical Name or Material Dichloro[1,1'-bis(dicyclohexylphosphino)ferrocene]palladium(II)
Physical Form Crystalline Solid
MDL Number MFCD11656081
CAS 917511-90-1
Synonym PdCl2(dcypf); [1,1'-Bis(dicyclohexylphosphino)ferrocene]dichloropalladium(II)
Concentration Pd 14.1%
Color Orange
Quantity 1g