missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II), Pd 16.3%

Coupling reactions, Suzuki couplings

$108.38 - $108.38

Chemical Identifiers

CAS 95408-45-0
Molecular Formula C26H44Cl2FeP2Pd
Formula Weight 651.77
MDL Number MFCD08064219
Synonym [1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride; PdCl2(dtbpf)
Chemical Name or Material Dichloro[1,1'-bis (di-tert-butylphosphino)ferrocene]palladium (II)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4482977 SDS Alfa Aesar™
4482977
0.1g Each for $108.38
AA4482901 SDS Alfa Aesar™
4482901
0.25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA4482903 SDS Alfa Aesar™
4482903
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

  • Packaged under Argon
Description & Specifications

Chemical Identifiers

95408-45-0
651.77
[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride; PdCl2(dtbpf)
C26H44Cl2FeP2Pd
MFCD08064219
Dichloro[1,1'-bis (di-tert-butylphosphino)ferrocene]palladium (II)

Specifications

95408-45-0
C26H44Cl2FeP2Pd
MFCD08064219
Dark red
0.1g
Crystalline
651.77
[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride; PdCl2(dtbpf)
Pd 16.3%
Dichloro[1,1'-bis (di-tert-butylphosphino)ferrocene]palladium (II)

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 :

  • Packaged under Argon