Please login to your online account to display your discounted pricing

Alfa Aesar™ Dicarbonylcyclopentadienylcobalt

Catalyst for the cyclotrimerization of alkynes

$151.00 - $151.00

Chemical Identifiers

Chemical Name or Material Dicarbonylcyclopentadienylcobalt
CAS 12078-25-0
Formula Weight 180.05
MDL Number MFCD00009589
UN Number UN3281
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2313606

  SDS
alfa aesar™
2313606*
5g Each for $151.00

AA2313603

  SDS
alfa aesar™
2313603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 180.05
Health Hazard 3 GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Dicarbonylcyclopentadienylcobalt
Physical Form Liquid
MDL Number MFCD00009589
CAS 12078-25-0
UN Number UN3281
Boiling Point 139° to 140°C (710mmHg)
Flash Point 26° (80°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H226-H341-H317
Fatal if swallowed.
Flammable liquid and vapor.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, LIQUID, N.O.S.
Sensitivity Air Sensitive
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H226-H341-H317
Fatal if swallowed.
Flammable liquid and vapor.
Suspected of causing genetic defects.
May cause an allergic skin reaction.