missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dicarbonylcyclopentadienylcobalt

Catalyst for the cyclotrimerization of alkynes

Manufacturer:  Alfa Aesar™ 2313603

 View more versions of this product

Catalog No. AA2313603



Specifications

Specifications

Dicarbonylcyclopentadienylcobalt
Air Sensitive
1g
MFCD00009589
180.05
26° (80°F)
139°C to 140°C (710mmHg)
12078-25-0
UN3281
Liquid

Chemical Identifiers

12078-25-0
MFCD00009589
Safety and Handling

Safety and Handling

GHS H Statement
H300-H226-H341-H317
Fatal if swallowed.
Flammable liquid and vapor.
Suspected of causing genetic defects.
May cause an allergic skin reaction.

GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, LIQUID, N.O.S.

SDS Documents