Please login to your online account to display your discounted pricing

Dicarbonyl(pentamethylcyclopentadienyl)chromium dimer, ACROS Organics™


Quantity 5g
Packaging Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  347970050

 View more versions of this product

Catalog No. AC347970050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystals
Formula Weight 486.49
Quantity 5g
Color Gray-black
Packaging Glass Bottle in Tin Can
Chemical Name or Material Dicarbonyl(pentamethylcyclopentadienyl)chromium dimer
Infrared Spectrum Authentic
Molecular Formula C24 H30 Cr2 O4
MDL Number MFCD00058843
CAS 37299-12-0
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement
Fatal in contact with skin.
Fatal if swallowed.
Fatal if inhaled.
Health Hazard 3 GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Dicarbonyl(pentamethylcyclopentadienyl)chromium dimer
CAS 37299-12-0
Molecular Formula C24 H30 Cr2 O4
Formula Weight 486.49
MDL Number MFCD00058843
Accessories

Danger

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Fatal in contact with skin.
Fatal if swallowed.
Fatal if inhaled.

SDS & Safety Documents