missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dicarbonyl(pentamethylcyclopentadienyl)chromium dimer, ACROS Organics™

Chemical Identifiers

CAS 37299-12-0
Molecular Formula C24 H30 Cr2 O4
Formula Weight 486.49
MDL Number MFCD00058843
Chemical Name or Material Dicarbonyl(pentamethylcyclopentadienyl)chromium dimer
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC347970010 SDS Thermo Fisher Scientific
347970010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC347970050 SDS Thermo Fisher Scientific
347970050
Glass Bottle in Tin Can 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

37299-12-0
486.49
Dicarbonyl(pentamethylcyclopentadienyl)chromium dimer
C24 H30 Cr2 O4
MFCD00058843

Specifications

486.49
1g
Glass bottle
C24 H30 Cr2 O4
MFCD00058843
Crystals
Gray-black
Authentic
37299-12-0
Dicarbonyl(pentamethylcyclopentadienyl)chromium dimer

Safety and Handling

GHS P Statement
Fatal in contact with skin.
Fatal if swallowed.
Fatal if inhaled.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger