Please login to your online account to display your discounted pricing

Dicarbonyl(2,4-pentanedionato)rhodium(I), Green crystalline, 99%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Dicarbonyl(2, 4-pentanedionato)rhodium(I)
CAS 14874-82-9
Physical Form Crystalline
Assay Percent Range 99%
Molecular Formula C7H7RhO4
Formula Weight 258.04
MDL Number MFCD00009884
Synonym (Acetylacetonato)dicarbonylrhodium(I)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3929503

  SDS
alfa aesar
3929503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3929506

  SDS
alfa aesar
3929506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA39295MD

  SDS
alfa aesar
39295MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Dicarbonyl(2, 4-pentanedionato)rhodium(I)
CAS 14874-82-9
Health Hazard 1 H301-H319-H317-H228-H412: Toxic if swallowed. Causes serious eye irritation. May cause an allergic skin reaction. Flammable solid. Harmful to aquatic life with long lasting effects.
DOT Information DOT Class: 4.1, Packing Group: II
EINECS Number 238-947-9
Odor Odorless
Physical Form Crystalline
Assay Percent Range 99%
Molecular Formula C7H7RhO4
Formula Weight 258.04
MDL Number MFCD00009884
Synonym (Acetylacetonato)dicarbonylrhodium(I)
Color Green
Quantity 1g
Safety and Handling

H301-H319-H317-H228-H412: Toxic if swallowed. Causes serious eye irritation. May cause an allergic skin reaction. Flammable solid. Harmful to aquatic life with long lasting effects.