Learn More
Dicalite™ speed plus, Diatomaceous Filter-aid, Acros Organics™
Manufacturer: Thermo Fisher Scientific 123380010
Description
Filter aidSpecifications
permeability flow ratio: 85 to 115 | |
White | |
9 to 10.5 (10% slurry) | |
< 1% | |
Plastic bottle | |
CNa2O3 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
disodium;carbonate | |
10340 | |
Powder |
Dicalite™ speed plus | |
1kg | |
68855-54-9 | |
Retained on 40 Mesh sieve: <0.90 wt.%, retained on 140 Mesh sieve: <10.0 wt.% | |
< 26.0 lbs/ft3 | |
MFCD00146688 | |
C(=O)([O-])[O-].[Na+].[Na+] | |
105.988 | |
CHEBI:29377 |
Chemical Identifiers
68855-54-9 | |
105.988 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
CHEBI:29377 | |
C(=O)([O-])[O-].[Na+].[Na+] |
CNa2O3 | |
MFCD00146688 | |
10340 | |
disodium;carbonate |
Safety and Handling
GHS H Statement:
May cause damage to organs through prolonged or repeated exposure if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning