missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dibutyltin dichloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC194870250

 View more versions of this product

Catalog No. AC194870250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
303.83
White to beige
01,213; 01,1294; 05,622
683-18-1
C8H18Cl2Sn
MFCD00000518
Glass bottle
Crystalline Low Melting Mass
25g
1.4g/mL
Dibutyltin dichloride
97%
Authentic
{CH3(CH2)3}2SnCl2
Dibutyldichlorotin

Chemical Identifiers

97%
{CH3(CH2)3}2SnCl2
MFCD00000518
Dibutyltin dichloride
C8H18Cl2Sn
303.83
Dibutyldichlorotin
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement
Very toxic to aquatic life with long lasting effects. Causes severe skin burns and eye damage. Suspected of causing genetic defects. Causes damage to organs through prolonged or repeated exposure. Harmful in contact with skin. Fatal if inhaled. Toxic if swallowed. May damage fertility. May damage the unborn child.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment.

Danger