Please login to your online account to display your discounted pricing

Dibutylboron trifluoromethanesulfonate, 1M solution in dichloromethane, AcroSeal™, ACROS Organics™

$104.03 - $245.32

Chemical Identifiers

Chemical Name or Material Dibutylboron trifluoromethanesulfonate
CAS 60669-69-4
CAS 60-29-7
Assay Percent Range 98%
Molecular Formula C9H18BF3O3S
Linear Formula CF3SO3B[(CH2)3CH3]2
Formula Weight 274.11
MDL Number MFCD00009669
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC378990250

 
thermo fisher scientific
AC378990250
Glass Bottle 25mL Each for $104.03

AC378991000

 
thermo fisher scientific
AC378991000
Glass Bottle 100mL Each for $245.32
Description & Specifications

Specifications

Physical Form Solution
Formula Weight 274.11
Quantity 25mL
Color Clear light yellow to orange
Packaging Glass Bottle
Density 0.8150g/mL
Fieser 07,91; 09,140; 16,109
Chemical Name or Material Dibutylboron trifluoromethanesulfonate
Assay Percent Range 98%
Molecular Formula C9H18BF3O3S
Linear Formula CF3SO3B[(CH2)3CH3]2
MDL Number MFCD00009669
CAS 60669-69-4
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Extremely flammable liquid and vapour. Harmful if swallowed. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. May form explosive peroxides. Repeated exposure may cause skin d
CAS 60-29-7
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsin

Lewis acid

Safety and Handling

Danger

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsin

GHS P Statement Extremely flammable liquid and vapour. Harmful if swallowed. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. May form explosive peroxides. Repeated exposure may cause skin d