missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dibutylamine (Reagent), Fisher Chemical

Manufacturer:  Thermo Scientific O2199500

 View more versions of this product

Catalog No. O2199-500




Specifications

Specifications

154.5° to 162.0°C
0.76g/cm³
Pass Test
1.4165 to 1.4185 (at 20°C)
159.6°C ± 0.1°C
-62°C
111-92-2
[CH3(CH2)3]2NH
2248
JQVDAXLFBXTEQA-UHFFFAOYSA-N
N-butylbutan-1-amine
8148
Liquid
Dibutylamine
47°C
Glass Bottle
2.59mmHg at 25°C
Undesignated
500mL
C8H19N
MFCD00009429
1-butanamine, n-butyl
CCCCNCCCC
129.247
129.25
Reagent

Chemical Identifiers

111-92-2
129.247
JQVDAXLFBXTEQA-UHFFFAOYSA-N
8148
CCCCNCCCC
C8H19N
MFCD00009429
1-butanamine, n-butyl
N-butylbutan-1-amine
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8, : Corrosive

SDS Documents