missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dibutylamine (Reagent), Fisher Chemical™
Supplier: Fisher Chemical O21991
Specifications
Dibutylamine | |
154.5° to 162.0°C | |
0.76g/cm³ | |
47°C | |
C8H19N | |
[CH3(CH2)3]2NH | |
1 L | |
dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine | |
CCCCNCCCC | |
129.247 | |
8148 | |
2.59mmHg at 25°C | |
Liquid |
111-92-2 | |
-62°C | |
159.6°C ± 0.1°C | |
Glass Bottle | |
1.4165 to 1.4185 (at 20°C) | |
MFCD00009429 | |
2248 | |
JQVDAXLFBXTEQA-UHFFFAOYSA-N | |
N-butylbutan-1-amine | |
4.46g/mol | |
129.25 | |
Reagent |
Chemical Identifiers
111-92-2 | |
129.247 | |
JQVDAXLFBXTEQA-UHFFFAOYSA-N | |
8148 | |
CCCCNCCCC |
C8H19N | |
MFCD00009429 | |
dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine | |
N-butylbutan-1-amine |
Safety and Handling
DOTInformation : DOT Class 8, : Corrosive