missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dibutyl succinate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC453010050

 View more versions of this product

Catalog No. AC453010050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Dibutyl succinate
144°C
274.0°C
5mL
98.5% min. (GC)
C4H9O2CH2CH2CO2C4H9
Solubility in water: slightly.
CCCCOC(=O)CCC(=O)OCCCC
230.3
CHEBI:82263
99%
0.9800g/mL
0.98
-29.0°C
141-03-7
C12H22O4
butanedioic acid, dibutyl ester, butyl butanedioate, di-n-butyl succinate, dibutyl succinate, dibutylsuccinate, dnbs, succinic acid di-n-butyl ester, succinic acid dibutyl ester, tabatrex, tabutrex
YUXIBTJKHLUKBD-UHFFFAOYSA-N
dibutyl butanedioate
8830
230.3

Chemical Identifiers

141-03-7
230.3
butanedioic acid, dibutyl ester, butyl butanedioate, di-n-butyl succinate, dibutyl succinate, dibutylsuccinate, dnbs, succinic acid di-n-butyl ester, succinic acid dibutyl ester, tabatrex, tabutrex
CHEBI:82263
CCCCOC(=O)CCC(=O)OCCCC
C12H22O4
YUXIBTJKHLUKBD-UHFFFAOYSA-N
8830
dibutyl butanedioate
Safety and Handling

Safety and Handling

EINECSNumber : 205-449-8

SDS Documents