Please login to your online account to display your discounted pricing

Dibutyl phthalate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Dibutyl phthalate
CAS 84-74-2
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C16H22O4
Formula Weight 278.34g/mol
MDL Number MFCD00009441
Synonym DBP
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC166600250

  SDS
thermo fisher scientific
166600250
Metal drum 25L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Free Acid 0.01% max.
Infrared Spectrum Authentic
Packaging Metal drum
Refractive Index 1.4910 to 1.4930 (20°C, 589nm)
Water 0.1% max.
Density 1.0430g/mL
Color Clear
Quantity 25L
Chemical Name or Material Dibutyl phthalate
CAS 84-74-2
Physical Form Clear liquid
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C16H22O4
Formula Weight 278.34g/mol
MDL Number MFCD00009441
Beilstein 09, II, 586
Merck Index 15, 3044
Synonym DBP
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May damage the unborn child. Suspected of damaging fertility.
Very toxic to aquatic life.
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.