Please login to your online account to display your discounted pricing

Dibutyl phthalate, 99%, ACROS Organics™


Quantity 25L
Packaging Metal drum

Manufacturer: thermo fisher scientific  166600250

 View more versions of this product

Catalog No. AC166600250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Free Acid 0.01% max.
Infrared Spectrum Authentic
Packaging Metal drum
Refractive Index 1.4910 to 1.4930 (20°C, 589nm)
Water 0.1% max.
Density 1.0430g/mL
Color Clear
Quantity 25L
Chemical Name or Material Dibutyl phthalate
CAS 84-74-2
Physical Form Clear liquid
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C16H22O4
Formula Weight 278.34g/mol
MDL Number MFCD00009441
Beilstein 09, II, 586
Merck Index 15, 3044
Synonym DBP
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May damage the unborn child. Suspected of damaging fertility.
Very toxic to aquatic life.
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Dibutyl phthalate
CAS 84-74-2
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C16H22O4
Formula Weight 278.34g/mol
MDL Number MFCD00009441
Synonym DBP
Accessories

Danger

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May damage the unborn child. Suspected of damaging fertility.
Very toxic to aquatic life.

SDS & Safety Documents