Learn More
Dibutyl phosphate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 406642500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dibutyl phosphate | |
107-66-4 | |
-13.0°C | |
250.0°C | |
Authentic | |
Glass bottle | |
1.4270 to 1.4290 | |
MFCD00015245 | |
1.058 | |
Solubility in water: 17.2g/L | |
CCCCOP(O)(=O)OCCCC | |
210.21 | |
75 mPa.s (20°C) | |
97% |
97% | |
259 to 267mg KOH/g | |
1.0580g/mL | |
180°C | |
97% | |
C8H19O4P | |
(CH3(CH2)3O)2P(O)OH | |
250 g | |
dibutyl phosphate, di-n-butyl phosphate, phosphoric acid, dibutyl ester, dibutyl acid phosphate, di-n-butylphosphoric acid, dp 4 coupling agent, di-n-butyl hydrogen phosphate, unii-0072nn74tn, phosphoric acid di-n-butyl ester, phosphoric acid dibutyl ester | |
JYFHYPJRHGVZDY-UHFFFAOYSA-N | |
dibutyl hydrogen phosphate | |
7881 | |
210.21 | |
Liquid |
Chemical Identifiers
107-66-4 | |
210.21 | |
JYFHYPJRHGVZDY-UHFFFAOYSA-N | |
7881 | |
CCCCOP(O)(=O)OCCCC |
C8H19O4P | |
MFCD00015245 | |
dibutyl phosphate, di-n-butyl phosphate, phosphoric acid, dibutyl ester, dibutyl acid phosphate, di-n-butylphosphoric acid, dp 4 coupling agent, di-n-butyl hydrogen phosphate, unii-0072nn74tn, phosphoric acid di-n-butyl ester, phosphoric acid dibutyl ester | |
dibutyl hydrogen phosphate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Suspected of causing cancer.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
EINECSNumber : 203-509-8
RTECSNumber : TB9605000
TSCA : TSCA
RUO – Research Use Only