missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dibutyl Malonate 99.0+%, TCI America™

Supplier:  TCI America M0540500G

 View more versions of this product

Catalog No. M0540500G



Description

Description

Specifications

Specifications

Dibutyl Malonate
Colorless
C11H20O4
500 g
NFKGQHYUYGYHIS-UHFFFAOYSA-N
1,3-dibutyl propanedioate
136925
≥99.0% (GC)
1190-39-2
251°C
MFCD00053795
Malonic Acid Dibutyl Ester
CCCCOC(=O)CC(=O)OCCCC
216.28
216.28
Liquid

Chemical Identifiers

1190-39-2
216.28
NFKGQHYUYGYHIS-UHFFFAOYSA-N
136925
CCCCOC(=O)CC(=O)OCCCC
C11H20O4
MFCD00053795
Malonic Acid Dibutyl Ester
1,3-dibutyl propanedioate
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions