Please login to your online account to display your discounted pricing

Dibutyl L-tartrate 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Dibutyl L-tartrate
Name Note 98%
CAS 87-92-3
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H22O6
Linear Formula CH3(CH2)3OCOCHOHCHOHCOO(CH2)3CH3
Formula Weight 262.29
MDL Number MFCD00009443
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC406660050

  SDS
thermo fisher scientific
406660050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid After Melting
Formula Weight 262.29
Quantity 5g
Color Clear colorless to yellow
EINECS Number 201-784-9
TSCA TSCA
Specific Rotation Condition + 10.50 (20.00°C c=neat)
Density 1.0900g/mL
Packaging Glass bottle
Refractive Index 1.4460 to 1.4480 (20°C, 589nm)
Specific Rotation +10° to +11° (20°C, 589nm) (neat)
Chemical Name or Material Dibutyl L-tartrate
Infrared Spectrum Authentic
Name Note 98%
Assay Percent Range 98%
Molecular Formula C12H22O6
Linear Formula CH3(CH2)3OCOCHOHCHOHCOO(CH2)3CH3
MDL Number MFCD00009443
CAS 87-92-3