missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dibutyl DL-Malate, TCI America™

$105.42 - $1,118.36

Chemical Identifiers

CAS 6280-99-5
Molecular Formula C12H22O5
Molecular Weight (g/mol) 246.303
MDL Number MFCD00059251
InChI Key PDSCSYLDRHAHOX-UHFFFAOYSA-N
Synonym DL-Apple Acid Dibutyl Ester, DL-Malic Acid Dibutyl Ester
PubChem CID 95385
IUPAC Name dibutyl 2-hydroxybutanedioate
SMILES CCCCOC(=O)CC(C(=O)OCCCC)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M002325G View Documents TCI America
M002325G
25g Each for $105.42
 
M0023500G View Documents TCI America
M0023500G
500g Each for $1,118.36
 
Specifications

Chemical Identifiers

6280-99-5
246.303
PDSCSYLDRHAHOX-UHFFFAOYSA-N
95385
CCCCOC(=O)CC(C(=O)OCCCC)O
C12H22O5
MFCD00059251
DL-Apple Acid Dibutyl Ester, DL-Malic Acid Dibutyl Ester
dibutyl 2-hydroxybutanedioate

Specifications

Yellow
6280-99-5
MFCD00059251
PDSCSYLDRHAHOX-UHFFFAOYSA-N
dibutyl 2-hydroxybutanedioate
95385
Clear Liquid at 20°C
Dibutyl DL-Malate
25g
C12H22O5
DL-Apple Acid Dibutyl Ester, DL-Malic Acid Dibutyl Ester
CCCCOC(=O)CC(C(=O)OCCCC)O
246.303
246.30
≥95.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1444

TSCA : Yes