missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dibucaine hydrochloride, Thermo Scientific Chemicals

Potent, long-acting local anesthetic

Supplier:  Thermo Scientific Chemicals J6280414

 View more versions of this product

Catalog No. AAJ6280414



Description

Description

It is applied as a potent, long-acting local anesthetic. It is used as an anaesthetic agent for spinal anaesthesia. More commonly it is a component in rectal medications used in inflammation of the anus and for the symptomatic treatment of haemorrhoids and fissures in the anus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as a potent, long-acting local anesthetic. It is used as an anaesthetic agent for spinal anaesthesia. More commonly it is a component in rectal medications used in inflammation of the anus and for the symptomatic treatment of haemorrhoids and fissures in the anus.

Solubility
Freely soluble in water, alcohol, acetone, or chloroform

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

Dibucaine hydrochloride
White
MFCD00012735
14,3031
Freely soluble in water, alcohol, acetone, or chloroform
CCCCOC1=NC2=CC=CC=C2C(=C1)C(=O)NCCN(CC)CC.Cl
379.929
CHEBI:59735
Crystalline Powder
61-12-1
C20H30ClN3O2
25 g
dibucaine hydrochloride, dibucaine hcl, cinchocaine hydrochloride, cinchocaine hcl, cincaine chloride, dibucaine cinchocaine hcl, percain, percaine, sovcain
IVHBBMHQKZBJEU-UHFFFAOYSA-N
2-butoxy-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide;hydrochloride
521951
379.92

Chemical Identifiers

61-12-1
379.929
IVHBBMHQKZBJEU-UHFFFAOYSA-N
521951
2-butoxy-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide;hydrochloride
C20H30ClN3O2
MFCD00012735
dibucaine hydrochloride, dibucaine hcl, cinchocaine hydrochloride, cinchocaine hcl, cincaine chloride, dibucaine cinchocaine hcl, percain, percaine, sovcain
CHEBI:59735
CCCCOC1=NC2=CC=CC=C2C(=C1)C(=O)NCCN(CC)CC.Cl
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.

P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c

H302-H318

EINECSNumber : 200-498-1

RTECSNumber : GD3325000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only