Learn More
Dibromomethane, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 112830025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Dibromomethane | |
74-95-3 | |
96.0°C to 98.0°C | |
98.5% min. (GC) | |
CH2Br2 | |
CH2Br2 | |
2.5L | |
11,337; 12,322; 16,393 | |
15,6133 | |
Solubility in water: 1g/L (20°C). Other solubilities: miscible with alcohol, ether, acetone and, chloroform | |
C(Br)Br | |
173.83 | |
CHEBI:47077 | |
99% |
99% | |
2.4700g/mL | |
Authentic | |
Glass Bottle | |
1.5400 to 1.5420 | |
MFCD00000168 | |
01,67 | |
2.47 | |
methylene bromide, methane, dibromo, methylene dibromide, dibromomethylene, rcra waste number u068, dibrommethan, methylenbromid, dibromo-methane, ccris 939, rcra waste no. u068 | |
FJBFPHVGVWTDIP-UHFFFAOYSA-N | |
dibromomethane | |
3024 | |
173.83 |
Safety and Handling
GHS H Statement
Harmful to aquatic life with long lasting effects.
Harmful if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 200-824-2
RTECSNumber : PA7350000
TSCA : TSCA
RUO â Research Use Only