missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dibromomethane, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 112835000

 View more versions of this product

Catalog No. AC112835000



Description

Description

Specifications

Specifications

Dibromomethane
Authentic
1.5400 to 1.5420
96.0°C to 98.0°C
-52.0°C
99%
98.5% min. (GC)
CH2Br2
01,67
15,6133
Solubility in water: 1g/L (20°C). Other solubilities: miscible with alcohol, ether, acetone and, chloroform
C(Br)Br
173.83
CHEBI:47077
Liquid
2.4700g/mL
Glass bottle
2.47
Colorless to Brown
500mL
74-95-3
CH2Br2
MFCD00000168
11,337; 12,322; 16,393
ccris 939, dibrommethan, dibromo-methane, dibromomethylene, methane, dibromo, methylenbromid, methylene bromide, methylene dibromide, rcra waste no. u068, rcra waste number u068
FJBFPHVGVWTDIP-UHFFFAOYSA-N
dibromomethane
3024
173.83
99%

Chemical Identifiers

74-95-3
173.83
FJBFPHVGVWTDIP-UHFFFAOYSA-N
3024
dibromomethane
CH2Br2
MFCD00000168
ccris 939, dibrommethan, dibromo-methane, dibromomethylene, methane, dibromo, methylenbromid, methylene bromide, methylene dibromide, rcra waste no. u068, rcra waste number u068
CHEBI:47077
C(Br)Br
Safety and Handling

Safety and Handling

GHS H Statement
Harmful to aquatic life with long lasting effects.
Harmful if inhaled.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 200-824-2

RTECSNumber : PA7350000

TSCA : TSCA

SDS Documents
Product Certifications