Please login to your online account to display your discounted pricing

Alfa Aesar™ Dibromobis(triphenylphosphine)nickel(II), 99%

As a polymerization catalyst

$24.18 - $224.73

Chemical Identifiers

Chemical Name or Material Dibromobis(triphenylphosphine)nickel(II)
CAS 14126-37-5
Formula Weight 743.12
MDL Number MFCD00015865
Synonym Bis(triphenylphosphine)nickel(II) bromide; Bis(triphenylphosphine)dibromonickel(II)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4219203

 
alfa aesar™
4219203
1g Each for $24.18

AA4219214

 
alfa aesar™
4219214
25g Each for $224.73
Description & Specifications

Specifications

Formula Weight 743.12
Chemical Name or Material Dibromobis(triphenylphosphine)nickel(II)
Melting Point 219° to 223°C
Physical Form Powder
Assay 99%
Quantity 1g
Odor Odorless
MDL Number MFCD00015865
CAS 14126-37-5
Synonym Bis(triphenylphosphine)nickel(II) bromide; Bis(triphenylphosphine)dibromonickel(II)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.