missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

alpha,alpha'-Dibromo-p-xylene, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 112940250

 View more versions of this product

Catalog No. AC112940250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

α,α′-Dibromo-p-xylene
623-24-5
100.0
White to Beige
245.0°C
96% min. (GC)
C8H8Br2
MFCD00000182
05, 385
1,4-bis bromomethyl benzene, p-xylylene dibromide, alpha,alpha'-dibromo-p-xylene, benzene, 1,4-bis bromomethyl, p-xylylene bromide, p-bromomethyl benzene, 1,4-dibromomethylbenzene, p-bis bromomethyl benzene, 1,4-bis-bromomethyl-benzene, ccris 1775
RBZMSGOBSOCYHR-UHFFFAOYSA-N
1,4-bis(bromomethyl)benzene
69335
97%
97%
96.0
142.0°C to 147.0°C
2.0120g/mL
Authentic
Glass bottle
C6H4(CH2Br)2
25 g
2.012
Solubility in water: soluble,hydrolyses
C1=CC(=CC=C1CBr)CBr
263.95
263.95
Crystalline Powder and Coarse Crystals

Chemical Identifiers

623-24-5
263.95
RBZMSGOBSOCYHR-UHFFFAOYSA-N
69335
C1=CC(=CC=C1CBr)CBr
C8H8Br2
MFCD00000182
1,4-bis bromomethyl benzene, p-xylylene dibromide, alpha,alpha'-dibromo-p-xylene, benzene, 1,4-bis bromomethyl, p-xylylene bromide, p-bromomethyl benzene, 1,4-dibromomethylbenzene, p-bis bromomethyl benzene, 1,4-bis-bromomethyl-benzene, ccris 1775
1,4-bis(bromomethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 210-781-1

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only