Please login to your online account to display your discounted pricing

Alfa Aesar™ Dibromo[2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]palladium(II)

Chemical Identifiers

Chemical Name or Material Dichloro[2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]palladium(II)
CAS 366488-99-5
Formula Weight 888.91
MDL Number MFCD14155710
Synonym PdBr2(BINAP); [2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl]dibromopalladium(II)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4666403

 
alfa aesar™
4666403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 888.91
Chemical Name or Material Dichloro[2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]palladium(II)
Physical Form Crystalline Solid
MDL Number MFCD14155710
CAS 366488-99-5
Synonym PdBr2(BINAP); [2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl]dibromopalladium(II)
Color Orange
Quantity 1g