missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dibromo[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), Pd 12.9%, Alfa Aesar™

Manufacturer:  Alfa Aesar 4498006

 View more versions of this product

Catalog No. AA4498006



Specifications

Specifications

Dibromo[1,1'-bis (diphenylphosphino)ferrocene]palladium (II)
124268-93-5
MFCD09953448
Pd 12.9%
C1=CC=C(C=C1)P(C2=CC=CC=C2)[C]3[CH][CH][CH][CH]3.C1=CC=C(C=C1)P(C2=CC=CC=C2)[C]3[CH][CH][CH][CH]3.[Fe].Br[Pd]Br
820.619
820.62
5g
C34H28Br2FeP2Pd
dibromo 1,1'-bis diphenylphosphino ferrocene palladium ii, dibromo 1,1'-bis diphenylphosphino ferrocene palladium ii
ZHEJVWMCZVGKAJ-UHFFFAOYSA-L
cyclopentyl(diphenyl)phosphane;dibromopalladium;iron
117064756
Solid

Chemical Identifiers

124268-93-5
820.619
ZHEJVWMCZVGKAJ-UHFFFAOYSA-L
117064756
C1=CC=C(C=C1)P(C2=CC=CC=C2)[C]3[CH][CH][CH][CH]3.C1=CC=C(C=C1)P(C2=CC=CC=C2)[C]3[CH][CH][CH][CH]3.[Fe].Br[Pd]Br
C34H28Br2FeP2Pd
MFCD09953448
dibromo 1,1'-bis diphenylphosphino ferrocene palladium ii, dibromo 1,1'-bis diphenylphosphino ferrocene palladium ii
cyclopentyl(diphenyl)phosphane;dibromopalladium;iron
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents