Please login to your online account to display your discounted pricing

Alfa Aesar™ Dibromo[1,1'-bis(diisopropylphosphino)ferrocene]palladium(II)

Chemical Identifiers

Chemical Name or Material Dibromo[1,1'-bis(diisopropylphosphino)ferrocene]palladium(II)
CAS 1154043-20-5
Formula Weight 684.55
MDL Number MFCD14155728
Synonym PdBr2(dippf); [1,1'-Bis(diisopropylphosphino)ferrocene]dibromopalladium(II)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4669603

 
alfa aesar™
4669603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Dibromo[1,1'-bis(diisopropylphosphino)ferrocene]palladium(II)
Color Red
Quantity 1g
CAS 1154043-20-5
Physical Form Crystalline Solid
Formula Weight 684.55
MDL Number MFCD14155728
Synonym PdBr2(dippf); [1,1'-Bis(diisopropylphosphino)ferrocene]dibromopalladium(II)