Please login to your online account to display your discounted pricing

Alfa Aesar™ Dibromo[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II)

Chemical Identifiers

Chemical Name or Material Dibromo[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II)
CAS 1000310-63-3
Formula Weight 740.66
MDL Number MFCD14155687
Synonym PdBr2(dtbpf); [1,1'-Bis(di-tert-butylphosphino)ferrocene]dibromopalladium(II)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4666903

  SDS
alfa aesar™
4666903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 740.66
Physical Form Crystalline Solid
Quantity 1g
MDL Number MFCD14155687
Color Dark brown
Sensitivity Air Sensitive
Synonym PdBr2(dtbpf); [1,1'-Bis(di-tert-butylphosphino)ferrocene]dibromopalladium(II)
CAS 1000310-63-3
Chemical Name or Material Dibromo[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II)