Please login to your online account to display your discounted pricing

Alfa Aesar™ Dibenzyl sulfone, 98+%

$41.14 - $135.16

Chemical Identifiers

Chemical Name or Material Dibenzyl sulfone
CAS 620-32-6
Molecular Formula C14H14O2S
Formula Weight 246.32
MDL Number MFCD00022036
Beilstein 2050498
Synonym Benzyl sulfone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0410806

  SDS
alfa aesar™
L0410806
5g Each for $41.14

AAL0410814

  SDS
alfa aesar™
L0410814
25g Each for $135.16
Description & Specifications

Specifications

Formula Weight 246.32
Chemical Name or Material Dibenzyl sulfone
MDL Number MFCD00022036
CAS 620-32-6
Synonym Benzyl sulfone
Molecular Formula C14H14O2S
Quantity 5g
Odor Odorless
Melting Point 149° to 152°C
Beilstein 2050498
Assay ≥98%