Please login to your online account to display your discounted pricing

Alfa Aesar™ Dibenzyl sulfide, 99%

$41.24 - $90.05

Chemical Identifiers

Chemical Name or Material Dibenzyl sulfide
CAS 538-74-9
Molecular Formula (C6H5CH2)2S
Formula Weight 214.33
MDL Number MFCD00004784
Beilstein 1911157
Merck Index 14,1145
Synonym Benzyl sulfide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2233714

 
alfa aesar™
B2233714
25g Each for $41.24

AAB2233722

 
alfa aesar™
B2233722
100g Each for $90.05
Description & Specifications

Specifications

Formula Weight 214.33
Chemical Name or Material Dibenzyl sulfide
TSCA TSCA
Melting Point 47° to 50°C
Assay 99%
Quantity 25g
Flash Point >110°C (230°F)
Molecular Formula (C6H5CH2)2S
MDL Number MFCD00004784
CAS 538-74-9
Beilstein 1911157
Merck Index 14,1145
Synonym Benzyl sulfide
RTECS Number DC0512500