Please login to your online account to display your discounted pricing

Alfa Aesar™ Dibenzyl phthalate, 97%

$103.35 - $396.28

Chemical Identifiers

Chemical Name or Material Dibenzyl phthalate
CAS 523-31-9
Molecular Formula C22H18O4
Formula Weight 346.39
MDL Number MFCD00014437
Beilstein 2338250
Synonym Phthalic acid dibenzyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0498506

  SDS
alfa aesar™
L0498506
5g Each for $103.35

AAL0498514

  SDS
alfa aesar™
L0498514
25g Each for $396.28
Description & Specifications

Specifications

Formula Weight 346.39
Chemical Name or Material Dibenzyl phthalate
CAS 523-31-9
Assay 97%
MDL Number MFCD00014437
Molecular Formula C22H18O4
Synonym Phthalic acid dibenzyl ester
Beilstein 2338250
Melting Point 40° to 42°C
Quantity 5g
Boiling Point 276° to 278°C (15mmHg)

Applications
Dibenzyl phthalate is an plasticizer used in toys that was found to exhibit potential estrogen agonist/ antagonist properties. It is used in PVC, paints, acrylic coatings.

Solubility
Insoluble in water.

Notes
Incompatible materials are reducing agents. Strong acids. Strong bases and Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.