missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dibenzyl phosphate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 112650250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dibenzyl phosphate | |
1623-08-1 | |
White to Yellow | |
98% | |
C14H15O4P | |
MFCD00004775 | |
06, 439 | |
HDFFVHSMHLDSLO-UHFFFAOYSA-N | |
dibenzyl hydrogen phosphate | |
74189 | |
98% |
99% | |
76.0°C to 80.0°C | |
Authentic | |
Glass bottle | |
(C6H5CH2O)2P(O)OH | |
25 g | |
dibenzyl phosphate, phosphoric acid dibenzyl ester, dibenzylphosphoric acid, unii-y65r35lz0s, phosphoric acid, bis phenylmethyl ester, dibenzylphosphate, creatine phosphate disodium impurity 4, dibenzyloxyphosphinic acid, dibenzyl-phosphate, pubchem14883 | |
C1=CC=C(C=C1)COP(=O)(O)OCC2=CC=CC=C2 | |
278.23 | |
278.23 | |
Powder |
Chemical Identifiers
1623-08-1 | |
278.23 | |
HDFFVHSMHLDSLO-UHFFFAOYSA-N | |
74189 | |
C1=CC=C(C=C1)COP(=O)(O)OCC2=CC=CC=C2 |
C14H15O4P | |
MFCD00004775 | |
dibenzyl phosphate, phosphoric acid dibenzyl ester, dibenzylphosphoric acid, unii-y65r35lz0s, phosphoric acid, bis phenylmethyl ester, dibenzylphosphate, creatine phosphate disodium impurity 4, dibenzyloxyphosphinic acid, dibenzyl-phosphate, pubchem14883 | |
dibenzyl hydrogen phosphate |
Safety and Handling
EINECSNumber : 216-602-3
RUO – Research Use Only