missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dibenzyl oxalate, 97%, ACROS Organics™

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C16H14O4
Linear Formula C6H5CH2OCOCOOCH2C6H5
Formula Weight 270.28
MDL Number MFCD00426620
Chemical Name or Material Dibenzyl oxalate
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC378400250 SDS Thermo Fisher Scientific
AC378400250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
C6H5CH2OCOCOOCH2C6H5
MFCD00426620
≥96.0%
C16H14O4
270.28
Dibenzyl oxalate

Specifications

7579-36-4
Glass bottle
Flaked Crystalline Solid
97%
C16H14O4
270.28
Dibenzyl oxalate
Authentic
75° to 78°C
White
25g
C6H5CH2OCOCOOCH2C6H5
MFCD00426620
≥96.0%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.