Please login to your online account to display your discounted pricing

Dibenzyl malonate, 90%, tech., ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: fisher scientific acros  170880500

 View more versions of this product

Catalog No. AC170880500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥88%
Formula Weight 284.3
Purity Grade Technical Grade
Physical Form Liquid
Quantity 50g
Color Clear yellow
EINECS Number 239-099-2
Density 1.1370g/mL
Beilstein 06, 436
Fieser 01,198
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5425 to 1.5465 (20°C, 589nm)
MDL Number MFCD00004779
Name Note tech., 90%
Linear Formula CH2(CO2CH2C6H5)2
Molecular Formula C17H16O4
CAS 15014-25-2
Chemical Name or Material Dibenzyl malonate
Assay Percent Range 90%

Specifications

Chemical Name or Material Dibenzyl malonate
Name Note tech., 90%
CAS 15014-25-2
Purity Grade Technical Grade
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C17H16O4
Linear Formula CH2(CO2CH2C6H5)2
Formula Weight 284.3
MDL Number MFCD00004779