Please login to your online account to display your discounted pricing

Dibenzyl malonate, 90%, tech., ACROS Organics™

Chemical Identifiers

Chemical Name or Material Dibenzyl malonate
Name Note tech., 90%
CAS 15014-25-2
Purity Grade Technical Grade
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C17H16O4
Linear Formula CH2(CO2CH2C6H5)2
Formula Weight 284.3
MDL Number MFCD00004779
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC170880500

  SDS
fisher scientific acros
170880500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Technical Grade
Physical Form Liquid
Assay ≥88%
Formula Weight 284.3
Quantity 50g
Color Clear yellow
EINECS Number 239-099-2
Density 1.1370g/mL
Beilstein 06, 436
Fieser 01,198
Chemical Name or Material Dibenzyl malonate
Refractive Index 1.5425 to 1.5465 (20°C, 589nm)
Name Note tech., 90%
Assay Percent Range 90%
Molecular Formula C17H16O4
Linear Formula CH2(CO2CH2C6H5)2
MDL Number MFCD00004779
CAS 15014-25-2
Infrared Spectrum Authentic
Packaging Glass bottle