missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dibenzyl Disulfide, TCI America™

$58.06 - $187.92

Chemical Identifiers

CAS 150-60-7
Molecular Formula C14H14S2
Molecular Weight (g/mol) 246.386
MDL Number MFCD00004783
InChI Key GVPWHKZIJBODOX-UHFFFAOYSA-N
Synonym dibenzyl disulfide, benzyl disulfide, disulfide, bis phenylmethyl, benzyl bisulfide, 1,2-dibenzyldisulfane, dibenzyl disulphide, dibenzyldisulfid, bis phenylmethyl disulfide, di phenylmethyl disulfide, disulfide, dibenzyl
PubChem CID 9012
ChEBI CHEBI:72752
IUPAC Name (benzyldisulfanyl)methylbenzene
SMILES C1=CC=C(C=C1)CSSCC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B041725G View Documents TCI America
B041725G
25g Each for $58.06
 
B0417500G View Documents TCI America
B0417500G
500g Each for $187.92
 
Specifications

Chemical Identifiers

150-60-7
246.386
GVPWHKZIJBODOX-UHFFFAOYSA-N
9012
(benzyldisulfanyl)methylbenzene
C14H14S2
MFCD00004783
dibenzyl disulfide, benzyl disulfide, disulfide, bis phenylmethyl, benzyl bisulfide, 1,2-dibenzyldisulfane, dibenzyl disulphide, dibenzyldisulfid, bis phenylmethyl disulfide, di phenylmethyl disulfide, disulfide, dibenzyl
CHEBI:72752
C1=CC=C(C=C1)CSSCC2=CC=CC=C2

Specifications

White-Yellow
25g
C14H14S2
dibenzyl disulfide, benzyl disulfide, disulfide, bis phenylmethyl, benzyl bisulfide, 1,2-dibenzyldisulfane, dibenzyl disulphide, dibenzyldisulfid, bis phenylmethyl disulfide, di phenylmethyl disulfide, disulfide, dibenzyl
C1=CC=C(C=C1)CSSCC2=CC=CC=C2
246.386
CHEBI:72752
Crystal-Powder at 20°C
Dibenzyl Disulfide
72°C
150-60-7
MFCD00004783
GVPWHKZIJBODOX-UHFFFAOYSA-N
(benzyldisulfanyl)methylbenzene
9012
246.39
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1136

RTECSNumber : JO1750000

TSCA : Yes