Please login to your online account to display your discounted pricing

Alfa Aesar™ Dibenzyl diethylphosphoramidite, 90+%

$114.96 - $390.26

Chemical Identifiers

Chemical Name or Material Dibenzyl diethylphosphoramidite
CAS 67746-43-4
Molecular Formula C18H24NO2P
Formula Weight 317.37
MDL Number MFCD00191987
Synonym DDP
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2561903

  SDS
alfa aesar™
B2561903
1g Each for $114.96

AAB2561906

  SDS
alfa aesar™
B2561906
5g Each for $390.26
Description & Specifications

Specifications

Formula Weight 317.37
Health Hazard 1 Warning
Assay ≥90%
Molecular Formula C18H24NO2P
Quantity 1g
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Flash Point 73°C (163°F)
Synonym DDP
CAS 67746-43-4
Chemical Name or Material Dibenzyl diethylphosphoramidite
MDL Number MFCD00191987
Recommended Storage 2° to 8°C

Applications
Dibenzyl N,N-diethylphosphoramidite may be used as phosphitylating reagent in the synthesis of glycosyl phosphites and phosphates, in the preparation of 6-dibenzylphosphate-1,3,5-tri-O-PMB-myo-inositol, for phosphorylating the alcohols.

Solubility
Soluble in chloroform.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

2° to 8°C