Learn More
Dibenzyl azodicarboxylate, 94%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 270270250
Description
C-X Bond Formation (Halogen), Radical initiatorsSpecifications
Dibenzyl azodicarboxylate | |
Authentic | |
Glass bottle | |
44°C to 52°C | |
94% | |
94% | |
MFCD00016737 | |
1,2-diazenedicarboxylic acid, 1,2-bis phenylmethyl ester, azodicarboxylic acid dibenzyl ester, dbad, diazenedicarboxylic acid, bis phenylmethyl ester, dibenzyl azodicarboxylate, dibenzyl diazene-1,2-dicarboxylate, dibenzylazodicarboxylate, o9nge39q8o, phenylmethyl benzyloxycarbonyl diazenyl formate, unii-o9nge39q8o | |
IRJKSAIGIYODAN-ISLYRVAYSA-N | |
benzyl (NE)-N-phenylmethoxycarbonyliminocarbamate | |
5387121 | |
Crystalline Powder |
>230°C | |
0.75% max. | |
Yellow to Orange | |
25g | |
2449-05-0 | |
C16H14N2O4 | |
15,111 | |
Solubility in water: insoluble | |
C1=CC=C(C=C1)COC(=O)N=NC(=O)OCC2=CC=CC=C2 | |
298.29 | |
298.29 | |
94% |
Chemical Identifiers
2449-05-0 | |
298.29 | |
IRJKSAIGIYODAN-ISLYRVAYSA-N | |
5387121 | |
C1=CC=C(C=C1)COC(=O)N=NC(=O)OCC2=CC=CC=C2 |
C16H14N2O4 | |
MFCD00016737 | |
1,2-diazenedicarboxylic acid, 1,2-bis phenylmethyl ester, azodicarboxylic acid dibenzyl ester, dbad, diazenedicarboxylic acid, bis phenylmethyl ester, dibenzyl azodicarboxylate, dibenzyl diazene-1,2-dicarboxylate, dibenzylazodicarboxylate, o9nge39q8o, phenylmethyl benzyloxycarbonyl diazenyl formate, unii-o9nge39q8o | |
benzyl (NE)-N-phenylmethoxycarbonyliminocarbamate |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable solid.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep container tightly closed.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 219-508-0
TSCA : TSCA