Please login to your online account to display your discounted pricing

Alfa Aesar™ Dibenzyl 5-aminoisophthalate, 96%

$62.34 - $190.86

Chemical Identifiers

Chemical Name or Material Dibenzyl 5-aminoisophthalate
CAS 152699-63-3
Molecular Formula C22H19NO4
Formula Weight 361.4
MDL Number MFCD03093065
Beilstein 744276
Synonym 5-Aminobenzene-1,3-dicarboxylic acid dibenzyl ester; 5-Aminoisophthalic acid dibenzyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1462903

  SDS
alfa aesar™
L1462903
1g Each for $62.34

AAL1462906

  SDS
alfa aesar™
L1462906
5g Each for $190.86
Description & Specifications

Specifications

Formula Weight 361.4
MDL Number MFCD03093065
Melting Point 116° to 118°C
Chemical Name or Material Dibenzyl 5-aminoisophthalate
Assay 96%
Molecular Formula C22H19NO4
CAS 152699-63-3
Beilstein 744276
Quantity 1g
Synonym 5-Aminobenzene-1,3-dicarboxylic acid dibenzyl ester; 5-Aminoisophthalic acid dibenzyl ester