missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dibenzoylmethane, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 112600250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Dibenzoylmethane | |
120-46-7 | |
Authentic | |
Plastic bottle | |
C6H5COCH2COC6H5 | |
25g | |
15,3014 | |
NZZIMKJIVMHWJC-UHFFFAOYSA-N | |
1,3-diphenylpropane-1,3-dione | |
8433 | |
224.26 |
98% | |
219°C to 221°C (18.0mmHg) | |
98% | |
C15H12O2 | |
MFCD00003085 | |
07,769 | |
dibenzoylmethane, 1,3-diphenyl-1,3-propanedione, 2-benzoylacetophenone, phenyl phenacyl ketone, 1,3-propanedione, 1,3-diphenyl, rhodiastab 83, omega-benzoylacetophenone, dibenzoyl-methane, karenzu dk2, unii-ans7me8okc | |
C1=CC=C(C=C1)C(=O)CC(=O)C2=CC=CC=C2 | |
224.26 | |
CHEBI:75417 | |
98% |
Safety and Handling
EINECSNumber : 204-398-9
RTECSNumber : TZ1930000
TSCA : TSCA
RUO â Research Use Only