missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(-)-Dibenzoyl-L-tartaric Acid Monohydrate 98%, ACROS Organics™

Manufacturer:  Acros Organics 160255000

 View more versions of this product

Catalog No. AC160255000



Specifications

Specifications

(-)-Dibenzoyl-L-tartaric acid monohydrate
Plastic bottle
-113° to -117° (20°C, 589nm) (c=5, ethanol)
White
98%
97.5
98%
[C6H5CO2CH(CO2H)-]2·H2O
2r,3r-2,3-bis benzoyloxy succinic acid hydrate, --dibenzoyl-l-tartaric acid monohydrate, --dibenzoyl-l-tartaric acidmonohydrate, dibenzoyl-l---tartaric acid monohydrate, l--dibenzoyl-l-tartatic acid monohydrate, 2r,3r---dibenzoyl-l-tartaric acid monohydrate, 2r,3r-2,3-bis benzoyloxy butanedioic acid hydrate, dibenzoyl-l-tartaric acid monohydrate, l-dbta, pubchem7152
C1=CC=C(C=C1)C(=O)OC(C(C(=O)O)OC(=O)C2=CC=CC=C2)C(=O)O.O
376.317
376.33
≥97.5% (on dry substance)
Authentic
−115° (20°C c=5,ethanol)
4 to 5.5%
500g
62708-56-9,2743-38-6
100.0
C18H16O9
MFCD00149119
DXDIHODZARUBLA-KAECKJJSSA-N
(2R,3S)-2,3-dibenzoyloxybutanedioic acid;hydrate
44119738
Powder

Chemical Identifiers

62708-56-9
376.317
DXDIHODZARUBLA-KAECKJJSSA-N
44119738
C1=CC=C(C=C1)C(=O)OC(C(C(=O)O)OC(=O)C2=CC=CC=C2)C(=O)O.O
C18H16O9
MFCD00149119
2r,3r-2,3-bis benzoyloxy succinic acid hydrate, --dibenzoyl-l-tartaric acid monohydrate, --dibenzoyl-l-tartaric acidmonohydrate, dibenzoyl-l---tartaric acid monohydrate, l--dibenzoyl-l-tartatic acid monohydrate, 2r,3r---dibenzoyl-l-tartaric acid monohydrate, 2r,3r-2,3-bis benzoyloxy butanedioic acid hydrate, dibenzoyl-l-tartaric acid monohydrate, l-dbta, pubchem7152
(2R,3S)-2,3-dibenzoyloxybutanedioic acid;hydrate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA

SDS Documents